Learn More
Trajan Scientific SYR 25ML 25MDF-LL-GT
SDP
Supplier:  Trajan Scientific 009463
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 25.0 mL, Diameter Outer: 1.06 in., 27 mm, Model: 25MDF-LL-GT, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-255-0764