Learn More
Trajan Scientific SYR 500UL 500F-GT-LT
SDP
Supplier:  Trajan Scientific 007229
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Tip Models, Volume: 500 uL, Diameter Outer: 0.31 in., 8 mm, Needle Gauge: No Needle Included, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone
Catalog No. 50-253-0316