Learn More
Med Vet International Dynarex Tuberculin Non-Safety Syringe - Luer Slip, 27g, 1/2" Needle, 1 Cc, 1000/Case
Supplier:  Med Vet International DYN6939
Encompass_Preferred
Dynarex Tuberculin Syringes are designed to help deliver accurate medication doses. The Syringes feature a double-beveled thin-walled needle for smoother insertion and improved patient comfort. The Tuberculin Syringes are constructed with securely attached needles to help prevent needle pop-off for increased safety. The Tuberculin Syringes feature removable plastic caps to help reduce the risk of accidental needlesticks for added protection. The Syringes have easy-to-read clear barrels with large font gradations to allow for clear visualization and precise measurement. Dynarex Tuberculin Syringes are sterile individually wrapped and sealed for optimal hygiene. Dynarex Tuberculin Syringes are high-quality syringes designed to deliver small doses of medication effectively and precisely. Each disposable Syringe features a sterile stainless-steel needle with a double-beveled tip for improved patient comfort.
Catalog No. 50-283-1579
 Restricted Item