Learn More
Medchemexpress LLC Syringaldehyde | 134-96-3 | 182.18 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN13905G
Encompass_Preferred
Syringaldehyde is a polyphenolic compound classified as a flavonoid. It is naturally found in various plant species, including *Manihot esculenta* and *Magnolia officinalis*. This compound has been observed to moderately inhibit COX-2 activity with an IC50 of 3.5 μg/mL. Additionally, it exhibits anti-hyperglycemic and anti-inflammatory properties.
  • Polyphenolic compound and flavonoid.
  • Found in plants such as *Manihot esculenta* and *Magnolia officinalis*.
  • Moderately inhibits COX-2 activity (IC50 of 3.5 μg/mL).
  • Exhibits anti-hyperglycemic activity.
  • Exhibits anti-inflammatory activity.
Catalog No. 50-004-14744