Learn More
Medchemexpress LLC (+)-Syringaresinol | 21453-69-0 | 98.7% | 418.44 g/mol | C22H26O8 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1260301MG
Encompass_Preferred
(+)-Syringaresinol is a lignan research compound that inhibits the Nuclear Factor of Activated T cells (NFAT) transcription factor, reported with an IC50 of 329.4 μM. It is provided as a high-purity material intended for in vitro research applications, including studies of lymphocytic leukemia and related cellular assays.
  • Inhibits NFAT transcription factor (IC50 329.4 μM).
  • Suitable for research of lymphocytic leukemia and related cellular studies.
  • High purity, 98.69%.
  • Molecular weight 418.44 g/mol; molecular formula C22H26O8.
  • Available in small research quantities (1 mg, 5 mg; larger sizes available by quote).
Catalog No. 50-002-36466