Learn More
Medchemexpress LLC (-)-Syringaresnol-4-O-β-D-apiofuranosyl-(1→2)-β-D-glucopyranoside | 136997-64-3 | 99.7% | 712.69 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN033810MM/1ML
Encompass_Preferred
(-)-Syringaresnol-4-O-β-D-apiofuranosyl-(1→2)-β-D-glucopyranoside is a lignan glycoside isolated from the stem bark of Albizia julibrissin Durazz. It is the main component of Albiziae cortex, which exhibits antidepressant, sedative, blood circulation promoting, and anti-edematous activities. This compound also shows potential anti-fertility and anticancer effects.
  • Exhibits antidepressant activity.
  • Has sedative effects.
  • Promotes blood circulation.
  • Shows anti-edematous activities.
  • Demonstrates potential anti-fertility effects.
  • May have anticancer effects.
Catalog No. 50-004-27216