Learn More
Waters Corp Syringe, 12.5 mL, 3767 Series, for HPLC System
Supplier:  Waters Corp 700012879
Encompass_Preferred
This replacement syringe is specifically designed for use with the 3767 Sample Manager within High-Performance Liquid Chromatography (HPLC) systems.
  • Provides a 12.5 mL volume.
  • Designed for specific use with the 3767 Sample Manager.
  • Integral component for HPLC systems.
  • Recommended for annual replacement.
Catalog No. 50-239-2628