Learn More
Hach Company Syringe, luer slip tip, 2 cc, 25/pk
Supplier:  Hach Company 2228725
Encompass
Disposable syringes feature a luer slip tip suited for connection with syringe filters or filter holders. Marked graduations. 25/pk
Catalog No. 50-199-5581