Learn More
Med Vet International AHS Syringe & Needle, 3mL, Luer Lock, 23G X 1", Hypodermic, 1000/Case, AH03L2325
Supplier:  Med Vet International AH03L2325X10
Encompass_Preferred
AHS AH03L2325 3mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This AHS Needle is a 23G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/CS
Catalog No. 50-304-4317