Learn More
Thermo Scientific Syringe 500Ul Gastight Ea
Supplier:  Thermo Scientific 0030119016
Encompass
500 microliter gas-tight all-plastic syringe for precise fluid measurement & dispensing in analytical applications like GC, LC & GC-MS; smooth sliding plunger with tight seals ensures accuracy & repeatability; gastight design prevents sample evaporation & air ingress; Luer lock tip allows secure connections; clear barrel for visual volume confirmation
Catalog No. NC9759850