Learn More
Trajan Scientific SYRINGE AUTO 5F-PE-GT-0.63
SDP
Supplier:  Trajan Scientific 001957
Encompass_Preferred
Trajan Autosampler Syringes, Connection Type: Fixed, Volume: 5 uL, Needle Gauge: 23 G, For Use With: PerkinElmer autosystem, Gas chromatography, Length Needle: 70 mm, 2.7 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 2.75 in., 70 mm, Model: 50F-PE-0.63C, Needle Point Style: Cone
Catalog No. 50-253-0411