Learn More
Sarstedt Inc Syringe filter, Filtropur S, PES, pore size: 0.2 µm, for sterile filtration
Supplier:  Sarstedt Inc 83.1826.001
Encompass_Preferred
Syringe filtration unit Filtropur S 0.2 | membrane: PES | filtration surf.: 5.3 cm2 | pore size: 0.2 µm for sterile filtr. | inlet: Luer-Lock female | outlet: Luer-Lock male | sterile | non-pyrogenic/endotoxin-free | non-cytotoxic | 1 pc./paper/plastic pack.
Catalog No. 501692591