Learn More
Trajan Scientific Syringe Gas tight GT 1mL 1/EA
SDP
Supplier:  Trajan Scientific 008160
Encompass_Preferred
1 mL syringe with gas tight plunger and removable Luer Lock push-pull valve
Catalog No. 50-004-94484