Learn More
Braintree Scientific Syringe Kit - Large
SDP
Supplier:  Braintree Scientific PSYRKITLG
Encompass
Large Syringe Kit includes: (2) 20 mL NormJect Luer Lock Inert(2) 30 mL NormJect Luer Lock Inert(2) 50 mL NormJect Luer Lock Inert(6) Female Luer Barbs 0.125"ID (3) Male Luer Barbs 0.125"ID(20 ft.) PVC Tubing 0.125"ID x 0.188"OD **Sold for research purposes only and not for human use**
Catalog No. 50-195-5582