Learn More
Trajan Scientific Syringe Nanovolume Needle Kit Repair Kit 1/EA
SDP
Supplier:  Trajan Scientific 033772
Encompass_Preferred
Replacemenet needle and plunger for 0 5 L NanoVolume CTC Analytics and Thermo Scientific syringes with 5 cm 0 63 mm OD cone tipped needle
Catalog No. 50-004-94571