Learn More
WELCH MATERIALS INC Welchrom Syringe Filter, NYLON, 25mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Supplier:  WELCH MATERIALS INC S25NY045B10
Encompass_Preferred
Syringe Filter, NYLON, 25mm, 0.45μm, Prefilter, Double layers,100pcs/pk,10pk/cs.
Catalog No. NC2052763