Learn More
Sigma Aldrich Fine Chemicals Biosciences Syringe pressure caps ster
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z120960100EA
Encompass
Disposable. for use with Luer lock or regular Luer syring tips covers entire Luer skirt of syringe.
Catalog No. 50-290-0646