Learn More
MIRION TECHNOLOGIES CAPINTEC INC PET Syringe Shield
Supplier: MIRION TECHNOLOGIES CAPINTEC INC 007980
The Pro-Tec PET Syringe Shield reduces hand exposure from syringes containing 511 keV radionuclides. The barrel of the shield is constructed of .34" thick tungsten that attenuates FDG F-18 by 88%.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.