Learn More
Med Vet International FlipLock Safety Tuberculin Syringes, TB, Exchangeable Needle, 3ml, 23G x 1in, 100/Box
Supplier:  Med Vet International 822331
Encompass_Preferred
EasyTouch FlipLock Safety Syringes offer an easy and simple solution in caregiver protection. After injection, just flip the protective shield over the needle until it clicks and locks in place. Hands and fingers stay away from the exposed needle. Simple, smart and cost-effective. Easy to Use Protective Cover Shields Needle After Use Familiar Technology Requires Limited Training Multi-lock Safety System Latex Free
Catalog No. 50-264-3611