Learn More
Med Vet International Terumo Syringe, 10mL, Luer Lock, 100/Box, SS-10L
Supplier:  Med Vet International 10L
Encompass_Preferred
Terumo SS-10L 10mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 10mL syringe fits conventional and safety needles. This quantity is 100/BX
Catalog No. 50-209-2975