Learn More
Med Vet International Terumo Tuberculin Syringe & Needle, 1mL, Removable Needle, 26GX 3/8in, 100/BX, SS-01T2609
Supplier:  Med Vet International 01T2609
Encompass_Preferred
Terumo SS-01T2609 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The syringe contains a removable needle. This Terumo SS-01T2609 1mL Syringe & Needle Combo comes with a 26GX 3/8' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a tuberculin syringe. They are small syringes with fine needles used to administer medication under the skin and perform tuberculosis tests. This quantity is 100/BX
Catalog No. 50-209-2978