Learn More
Trajan Scientific SYRINGES 25MDR VLLMA GT
SDP
Supplier:  Trajan Scientific 009472
Encompass_Preferred
SGE Syringes with Valves, Volume: 25 mL, Borosilicate Glass, PTFE Plunger, Autoclavable: Autoclavable, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Termination: Luer-lock
Catalog No. 50-253-0292