Learn More
Medchemexpress LLC Syringetin-3-O-glucoside | 40039-49-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN81945MG
Encompass_Preferred
Syringetin-3-O-glucoside is a flavonol glycoside that exhibits relatively weak DPPH and ABTS radical scavenging activity. It is intended for research use only.
  • Flavonol glycoside
  • Exhibits weak DPPH and ABTS radical scavenging activity
  • Purity of 99.37%
  • Solid appearance, light yellow to yellow in color
  • Derived from Vitis vinifera cv. Zalema plants
  • Classified as a flavonoid, flavonol, phenol, and polyphenol
Catalog No. 50-004-19165