Learn More
Medchemexpress LLC HY-N4115 5mg , Syrosingopine CAS:84-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41155MG
Encompass_Preferred
HY-N4115 5mg Syrosingopine CAS: 84-36-6 Syrosingopine (Su 3118) is an orally active lactate transporters (MCT1/MCT4) dual inhibitor, which can reduce glycolysis and induce synthetic lethality in cancer cells when combine with metformin. Syrosingopine shows anti-hypertensive activity by depleting peripheral stores of norepinephrine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9834