Learn More
Selleck Chemical LLC Syrosingopine Su-3118-5mg
SDP
Supplier:  Selleck Chemical LLC S99075MG
Encompass_Preferred
Syrosingopine is a dual MCT1 and MCT4 inhibitor with IC50 values of 2500nM and 40 nM in HAP1 cells, respectively.
Catalog No. 50-238-9596