Learn More
World Precision Instrument System 1: EVOM Manual, HTS STX, 1 Plate
SDP
Supplier:  World Precision Instrument EVMSYS010101
Encompass
World Precision Instruments (WPI) is excited to introduce the new EVOM™ Manual High Throughput Screening System. The new EVOM™ Manual HTS System features WPI’s EVOM™ Manual, the gold standard for delivering stable and repeatable Trans Epithelial Electrical Resistance (TEER) measurements, qualitatively measuring cell monolayer health and quantitatively measuring cell confluence.
Catalog No. 50-252-1181