Learn More
CPC Scientific H-Ala-Val-Gln-Ser-Lys-Pro-Pro-Ser-Lys-Arg-Asp-Pro-Pro-Lys-Met-Gln-Thr-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MISC013B
Encompass
Peptide isolated from tomato leaves that induces the synthesis of wound-inducible proteinase inhibitor proteins.SEQUENCE: H-Ala-Val-Gln-Ser-Lys-Pro-Pro-Ser-Lys-Arg-Asp-Pro-Pro-Lys-Met-Gln-Thr-Asp-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: AVQSKPPSKRDPPKMQTDMOLECULAR FORMULA: C85H144N26O28S1MOLECULAR WEIGHT: 2010.30STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [137181-56-7]SYNONYMS: RESEARCH AREA: MiscellaneousREFERENCES:G. Pearce et al., Science, 253, 895 (1991)SKU(s): MISC-013A, MISC-013B
Catalog No. 50-210-9739