Learn More
Ace Glass, Inc. Cannon-Fenske viscometer, calibrated, size 400, 240-1200centistokes
SDP
Supplier:  Ace Glass, Inc. 798849
Encompass
Cannon-Fenske viscometer, calibrated, size 400, 240-1200centistokes
Catalog No. 50-975-863