Learn More
MSC HYFLEX HPPE/SPNDX C/R GLV
Supplier:  MSC 92992460
Encompass
Cut & Puncture Resistant Gloves; Coating Material: Nitrile ; Coating Coverage: Palm & Fingertips ; Glove Type: Cut-Resistant; Abrasion Resistant ; ANSI/ISEA Cut Resistance Level: A9 ; Primary Material: Intercept - HPPE, Nylon, Spandex, Steel, Graphene
Catalog No. 50-273-6420