Learn More
Medchemexpress LLC SZM679 | 3027645-52-6 | C27H18F5N3O5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151873100MG
Encompass_Preferred
SZM679 is a potent, orally active, and selective RIPK1 inhibitor. It reverses tumor necrosis factor-induced systemic inflammatory response and decreases Tau hyperphosphorylation, neuroinflammation, and RIPK1 phosphorylation levels. This compound is valuable for research, particularly in the study of Alzheimer's disease.
  • Potent and selective RIPK1 inhibitor (Kd 8.6 nM)
  • Reverses TNF-induced systemic inflammatory response
  • Decreases Tau hyperphosphorylation and neuroinflammation
  • Reduces RIPK1 phosphorylation levels
  • Protects against necroptosis
  • Inhibits expression of RIPK1 and blocks necrosome formation
  • Improves cognitive function and anxiety in AD models
  • Rescues brain structure damage
Catalog No. 50-003-73267