Learn More
Medchemexpress LLC T-1101 tosylate | 2250404-95-4 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY120356A10MM 1ML
Encompass_Preferred
T-1101 tosylate is the tosylate salt of T-1101, an orally active inhibitor of Hec1-Nek2 with reported anticancer activity. Supplied as a ready-to-use 10 mM solution in DMSO (1 ML) or as solid powder in multiple sizes for research applications.
  • Ready-to-use 10 mM solution in DMSO, 1 ML.
  • High purity: 98.9% (HPLC).
  • Molecular weight 665.80 and formula C31H31N5O6S3.
  • Available in small solid sizes; larger quantities available by request.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-33297