Learn More
Medchemexpress LLC T-448 | 1597426-53-3 | 98.4% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY122635A100MG
Encompass_Preferred
T-448 is an orally active, irreversible inhibitor of lysine-specific demethylase 1 (LSD1) for research use. It inhibits LSD1 with an IC50 of about 22 nM, is supplied as a solid with high purity, and is DMSO-soluble with recommended in vivo formulation protocols.
  • Irreversible LSD1 inhibitor with an IC50 of 22 nM.
  • High purity solid suitable for biochemical and cellular studies.
  • DMSO soluble (≈16.67 mg/mL) with recommended vehicle formulations for in vivo use.
  • Available in multiple pack sizes, including 100 MG.
  • Provided with supporting documentation such as SDS and COA.
Catalog No. 50-002-30180