Learn More
TARGETMOL CHEMICALS INC T-5224 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T541610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. T-5224 is a transcription factor c-Fos/AP-1 inhibitor which specifically inhibits the DNA binding activity of c-Fos/c-Jun without affecting other transcription factors. purity: 99%
Catalog No. 50-257-9770