Learn More
Medchemexpress LLC T-Boc-aminooxy-PEG2-azide | 252378-68-0 | ≥97.0% | C11H22N4O5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140431
Encompass_Preferred
T-Boc-Aminooxy-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-52755