Learn More
Medchemexpress LLC T-Boc-aminooxy-PEG5-azide | 2250216-95-4 | 95.0% | C17H34N4O8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140434100MG
Encompass_Preferred
T-Boc-Aminooxy-PEG5-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, enabling efficient conjugation with various molecules.
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Compatible with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Reacts with molecules containing DBCO or BCN groups
Catalog No. 50-003-52940