Learn More
Broadpharm t-Boc-N-amido-PEG12-NHS 500mg
Supplier:  Broadpharm BP28086500MG
Encompass
t-Boc-N-amido-PEG12-NHS ester is a PEG linker containing a Boc-protected amino group and an NHS ester group. The hydrophilic PEG spacer increases solubility in aqueous media. The NHS ester can be used to label the primary amines (-NH2) of proteins| amine-modified oligonucleotides| and other amine-containing molecules. The protected amine can be deprotected in acidic conditions. 500 MG
Catalog No. NC2941537