Learn More
Medchemexpress LLC T-Boc-N-amido-PEG15-Br | 3012589-40-8 | 98.0% | C37H74BrNO17 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14382950MG
Encompass_Preferred
t-Boc-N-amido-PEG15-Br is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research use only.
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed to selectively degrade target proteins
  • For research use only
Catalog No. 50-003-55009