Learn More
Medchemexpress LLC t-Boc-N-amido-PEG5-acetic acid | 890152-41-7 | MFCD31656900 | 99.9% | 395.45 g/mol | C17H33NO9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW19096250MG
Encompass_Preferred
t-Boc-N-amido-PEG5-acetic acid is a Boc-protected PEG5 linker terminating in an acetic acid functional group intended for use in targeted protein degrader (PROTAC) synthesis. It provides a flexible, water-soluble spacer and a masked amine for selective functionalization, facilitating linker optimization and efficient conjugation in medicinal chemistry workflows.
  • Boc-protected amine for controlled deprotection and downstream derivatization.
  • PEG5 spacer provides flexibility and improved aqueous solubility.
  • Terminal acetic acid enables standard amide coupling chemistries.
  • High purity supports reproducible coupling and characterization.
  • Available in laboratory-scale quantities suitable for synthesis optimization.
Catalog No. 50-002-80747