Learn More
Medchemexpress LLC t-Boc-N-amido-PEG5-Tos | 1404111-69-8 | MFCD34596623 | 96.0% | 491.60 | C22H37NO9S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW460275250MG
Encompass_Preferred
t-Boc-N-amido-PEG5-Tos is a Boc-protected PEG5 linker bearing a tosyl leaving group. The hydrophilic PEG spacer improves solubility of conjugates, the tosyl enables efficient nucleophilic substitution, and the Boc group can be removed under mild acidic conditions to expose a primary amine. The reagent is intended for synthetic chemistry and bioconjugation applications.
  • Hydrophilic PEG5 spacer improves aqueous solubility.
  • Tosyl leaving group facilitates nucleophilic substitution.
  • Boc-protected amine allows orthogonal deprotection strategies.
  • Suitable for linker construction, bioconjugation, and PROTAC synthesis.
  • Available in small research quantities for laboratory use.
Catalog No. 50-002-84991