Learn More
Medchemexpress LLC t-Boc-N-amido-PEG5-Tos | 1404111-69-8 | MFCD34596623 | >98.0% | 491.6 | C22H37NO9S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW460275100MG
Encompass_Preferred
t-Boc-N-amido-PEG5-Tos is a Boc-protected PEG5 linker bearing a tosylate leaving group and a Boc-protected amine. The hydrophilic PEG5 spacer improves aqueous solubility and provides a flexible handle for substitution and conjugation reactions, making this compound useful as a building block in medicinal chemistry, linker synthesis, and bioconjugation workflows.
  • Hydrophilic PEG5 spacer increases aqueous solubility.
  • Boc-protected amine permits controlled deprotection under acidic conditions.
  • Tosylate group acts as an efficient leaving group for nucleophilic substitution.
  • Useful as a linker or building block for conjugation and PEGylation.
  • Compatible with standard organic synthesis and bioconjugation protocols.
Catalog No. 50-002-84214