Learn More
Strem, An Ascensus Company CAS# 1660153-91-2. 5G. 2-(t-Butylphenylphosphino)-2',6'-dimethylamino-1,1'-biphenyl, 98% (t-Bu)PhCPhos
SDP
Supplier:  Strem, An Ascensus Company 1530105G
Encompass
CAS# 1660153-91-2. 5G. 2-(t-Butylphenylphosphino)-2',6'-dimethylamino-1,1'-biphenyl, 98% (t-Bu)PhCPhos. Molecular Formula: C26H33N2P. Molecular Weight: 404.53. Color/Form: white to off-white pwdr. Strem# 15-3010. www.strem.com/catalog/v3/15-3010/
Catalog No. 50-196-3525