Learn More
Enzo Life Sciences t-Butylhydroquinone (10g). CAS: 1948-33-0
SDP
Supplier:  Enzo Life Sciences ALX270061G010
Encompass_Preferred
Ca2+ ATPase inhibitor. Erk2 activator. Inhibitor of the endoplasmic reticulum Ca2+-ATPase. Activates extracellular signal-regulated protein kinase 2 (ERK2). Alternative name: tBHQ. Formula: C10H14O2. MW: 166.2. Purity: ≥97% (GC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: Ambient.
Catalog No. 50-200-8394