Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRECIGENOME T-CELL TARGETED ANTIBODY- 1ML
SDP

Supplier:  PRECIGENOME PGLNPEGF02CD3Q1ML

Encompass

  • Targets CD3+ T cells
  • Contains anti-human CD3 (OKT3) mAb
  • Payload includes EGFP mRNA (m1Ψ)
  • Utilizes ionizable lipid SM102
  • Formulated with DSPC, PEG-2000 DMG, and cholesterol
  • Stored in 10% sucrose in TBS
  • Appearance is clear and colorless
  • Concentration approximately 0.1 mg/mL
  • Recommended storage at 4°C
  • Stable for 6 months from manufacture date
  • Designed for T cell research, immunotherapy, and cell tracking
  • Research-grade quality
  • Delivery via lipid nanoparticle system
  • mRNA modified with N1-methyl-pseudouridine (m1Ψ)
  • Covalent conjugation method
  • Available as a premade format
  • Ships with ice packs
  • Customization options for targeting antibodies and formulations

Catalog No. NC3781971


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.