Learn More
Bellco Glass, Inc. T Connector Fitting Pack, Polyethylene, 1/8" HB x 1/8" HB to 1/8" HB, 10/PK
SDP
Supplier:  Bellco Glass, Inc. 452800275
Encompass_Preferred
T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene. Material: PolyethyleneColor: BlackLength: 1.17"OD: 0.145"ID: 0.075"
Catalog No. 50-255-0487