Learn More
Bellco Glass, Inc. T Connector Fitting Pack, Polyethylene, 3/4" HB x 3/4" HB to 3/4" HB, 100/PK
SDP
Supplier:  Bellco Glass, Inc. 452800284
Encompass_Preferred
T connectors are available in a variety of standard sizes. Our NPT threaded and barbed tees offer strength and durability. Made from Polyethylene. Material: PolyethyleneColor: BlackLength: 2.98"OD: 0.785"ID: 0.550"
Catalog No. 50-255-0478