Learn More
MilliporeSigma T DIAGNOSTICS153 SIMULFLUOR
Supplier:  MilliporeSigma 5243
Encompass
SimulFluor HSV1/2 Reagent - One 5 mL dropper vial containing two components specific for HSV-1 and HSV-2, protein stabilizer, Evans blue and 0.1% sodium azide (preservative).
Catalog No. 50-580-388