Learn More
UFP TECHNOLOGIES INC T-TUBE STRAIGHT 3FT LONG 1.5IN
Supplier:  UFP TECHNOLOGIES INC 101415
Encompass
UFP Technologies' straight, non-jacketed T-Tubes come in 3 ft sections. Item 101415 has a line OD of 1.50 inches.
Catalog No. NC1952009