Learn More
Medchemexpress LLC T01-1, an anticancer agent (camptothecin derivative) | 2356229-14-4 | 99.9% | 511.59 | 5 MG

Supplier: Medchemexpress LLC HY1481855MG
T01-1 is an anticancer agent, a camptothecin derivative, exhibiting good anti-proliferative activity. It is intended for research use only.
- Anticancer agent (camptothecin derivative)
- Exhibits good anti-proliferative activity
- Inhibits HCC1806 cell proliferation (EC50 of 6.7 nM)
- Store at -20°C, protect from light
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
- Soluble in DMSO at 25 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.