Learn More
Selleck Chemical LLC T0901317 25mg 293754-55-9
SDP
Supplier:  Selleck Chemical LLC S707625MG
Encompass_Preferred
T0901317 is a potent and selective agonist for both LXR and FXR, with EC50 of ~50 nM and 5 ?M, respectively. T0901317 is a dual inverse agonist of ROR? and ROR? with Ki of 132 nM and 51 nM, respectively. T0901317 significantly suppresses cell proliferation and induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4376