Learn More
Alkali Scientific T3-0248G MultiCycler 248 Multi
SDP
Supplier:  Alkali Scientific RS0248G
Encompass_Preferred
Alkali Scieintific T3-0248G is designed for laboratories wanting flexibility in their applications Alkali Scientifics T3-0248G MultiCyclerThermal Cyclers offer multiple independent cycling blocks in a single unit Dual or triple block models are available both offering extremely fast cycling profiles large touch screen interfaces self-adjusting heated lids gradient capabilities and an easy-to-use program wizard All programming features are accessed from the large 10 full color touch screen The screen can be angled towards the user when in use and then tucked away to save space A program wizard guides users through the simple protocol set up process Advanced programming features include gradient adjustable ramp rates and time increments and decrements For multi-user labs password protection prevents interference of an existing run A USB provides for unlimited program storage and easy transfer of protocols between MultiCycler machines The auto-adjusting heated lid prevents condens
Catalog No. 50-302-9661
Add to Cart
Request a Quote